Learn More
Fisherbrand™ Universal Sorbents
Universal, nonselective sorbents can fully saturate in as little as 20 seconds.
$121.32
Specifications
| Material | Polypropylene |
|---|
Alert:
Do not use particulate, socks, or pillows on hydrofluoric acid. Do not use pads, rolls, rugs, or mats on concentrated sulfuric acid and strong caustics, as fabric casings may degrade if contact is lengthy. Refer to Material Safety Data Sheet for details.
| Catalog Number | Mfr. No. | Capacity (English) | Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Type | Price | Quantity & Availability | ||||
|
172062B
|
Fisherbrand™
FBHAZSOC12 |
23 gal. | Universal Sorbent Sock |
Case of 12 Each for $121.32
|
|
||||
Description
Pads and Rolls
- Made of polypropylene that has been shredded, pulped, and air-blown through two strong layers of spunbonded film, then heat-sealed and dimpled for extra strength
- Tough, lint-free, soft, pliable, and highly absorbent
- Fold easily upon themselves to form multi-layer barrier
- Suited for leaks, spills, or drips and for routine maintenance—can be used on mild acids and caustics
- Absorb up to 20 times their weight
- Saturated pads resist tearing and sticking to surfaces, and can be wrung out and then reused or incinerated with less than 0.1% ash residue
- Ideal for spill containment and control
- Casings made of chemical-resistant, nonwoven polyester blend (with polyester thread) that enhances absorbency
- Absorb up to 14 times their weight
Compliance
Nonbiodegradable; eliminate free liquids, meeting U.S. EPA requirements for landfill disposal of nonregulated wastes and sludges. Particulate passes Paint Filter and TCLP tests. Handle disposal according to federal, state, and local laws.
Specifications
| Polypropylene |