missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ UniStore Wide Mouth Square Bottles PPCO
These wide mouth square bottles are are made of polypropylene copolymer PPCO bottles and are autoclavable.
Specifications
| Color | Translucent White |
|---|---|
| Material | Plastic |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S01881
|
United Scientific™
33609 |
1000 mL |
N/A
|
|
|||||
|
S01887
|
United Scientific™
33605 |
60 mL |
N/A
|
|
|||||
|
S01888
|
United Scientific™
33606 |
175 mL |
N/A
|
|
|||||
|
S01879
|
United Scientific™
33607 |
250 mL |
N/A
|
|
|||||
|
S01880
|
United Scientific™
33608 |
500 mL |
N/A
|
|
|||||
Description
UniStore™ Wide Mouth Square Bottles are available in different materials and sizes for multiple applications. The square design allows the bottles to nest together for more efficient storage, packing and shipping. The polypropylene copolymer (PPCO) bottles are autoclavable at 121°C. These bottles are leakproof and have a polypropylene cap.
- Square design for compact storage
- Better impact strength and durability than standard polypropylene (PP)
- Precision molded for consistent wall thickness with smooth interior and exterior surfaces
- FDA-21CFR compliant resin
- Autoclavable at 121°C
- Leakproof closure
- Available in 60 mL, 175 mL, 250 mL, 500 mL, and 1000 mL
Specifications
| Translucent White | |
| Non-sterile | |
| No | |
| Reagent Bottle |
| Plastic | |
| Yes | |
| Wide, Threaded | |
| Autoclavable |