missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ UniStore Wide Mouth Square Bottles PPCO

Catalog No. S01888
Change view
Click to view available options
Capacity (Metric):
60 mL
175 mL
250 mL
500 mL
1000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S01888 175 mL
S01881 1000 mL
S01887 60 mL
S01879 250 mL
S01880 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S01888 Supplier United Scientific™ Supplier No. 33606

These wide mouth square bottles are are made of polypropylene copolymer PPCO bottles and are autoclavable.

UniStore™ Wide Mouth Square Bottles are available in different materials and sizes for multiple applications. The square design allows the bottles to nest together for more efficient storage, packing and shipping. The polypropylene copolymer (PPCO) bottles are autoclavable at 121°C. These bottles are leakproof and have a polypropylene cap.

  • Square design for compact storage
  • Better impact strength and durability than standard polypropylene (PP)
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces
  • FDA-21CFR compliant resin
  • Autoclavable at 121°C
  • Leakproof closure
  • Available in 60 mL, 175 mL, 250 mL, 500 mL, and 1000 mL

Specifications

Capacity (Metric) 175 mL
Color Translucent White
Material Plastic
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Autoclavable Autoclavable
Capacity (English) 6 oz.
Product Type Reagent Bottle