missing translation for 'onlineSavingsMsg'
Learn More

Undecane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Undecane is obtained from the refining of petroleum. It is a component of gasoline and is generally used in organic synthesis, petroleum research and as a distillation chaser.

$140.45 - $558.03

Specifications

CAS 1120-21-4
Vapor Pressure <0.4 mmHg (20°C)
Vapor Density 5.4 (vs. air)
Density 0.74 g/mL (at 25°C)
Flash Point 62°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019912
View Documents
Merck Emd Millipore
940005ML
5 mL
Each for $140.45
Only null left
 
111018403
View Documents
Merck Emd Millipore
9400025ML
25 mL
Each for $558.03
Only null left
 
Description

Description

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

1120-21-4
5.4 (vs. air)
62°C
CH3(CH2)9 CH3
-26°C
n-Undecane; Hendecane
156.31
Analytical Standard
<0.4 mmHg (20°C)
0.74 g/mL (at 25°C)
Neat
MFCD00008959
196°C
Analytical Standard
≥99.8% (GC)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'