missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecane, ≥99.8% (GC), MilliporeSigma™ Supelco™
Undecane is obtained from the refining of petroleum. It is a component of gasoline and is generally used in organic synthesis, petroleum research and as a distillation chaser.
Supplier: Merck Emd Millipore 9400025ML
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal CareSpecifications
| 1120-21-4 | |
| 5.4 (vs. air) | |
| 62°C | |
| CH3(CH2)9 CH3 | |
| -26°C | |
| n-Undecane; Hendecane | |
| 25 mL | |
| ≥99.8% (GC) |
| <0.4 mmHg (20°C) | |
| 0.74 g/mL (at 25°C) | |
| Neat | |
| MFCD00008959 | |
| 196°C | |
| Analytical Standard | |
| 156.31 | |
| Analytical Standard |