missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals UNC2025, 95%

Catalog No. AC467881000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467881000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC467881000 Supplier Thermo Scientific Chemicals Supplier No. 467881000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1429881-91-3 | C28H40N6O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1429881-91-3
Molecular Formula C28H40N6O

Specifications

Assay Percent Range 95%
Quantity 100 mg
Formula Weight 476.66
Chemical Name or Material UNC2025
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

If skin irritation occurs: Get medical advice/attention.

If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only