missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ UNC 6934

Catalog No. 361063843
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361063843 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361063843 Supplier R&D Systems™ Supplier No. 7183/50*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Antagonizes PWWP1 interaction with nucleosomal H3K36me2, alters NSD2 nucleolar localization

UNC 6934 binds to the N-terminal proline-tryptophan-tryptophan-proline (PWWP) domain of nuclear receptor-binding SET domain-containing 2 (NSD2). UNC 6934 inhibits PWWP1 interaction with nucleosomal H3K36me2 (IC50 = 104 nM) by occupying the canonical H3K36me2-binding pocket of PWWP1. It selectively binds endogenous NSD2 in cells and causes accumulation of endogenous NSD2 in the nucleolus.

Specifications

Chemical Name or Material N-Cyclopropyl-3,4-dihydro-3-oxo-N-[[4-[(4-pyrimidinylamino)carbonyl]phenyl]methyl]-2H-1,4-benzoxazine-7-carboxamide
CAS 2561494-77-5
Quantity 50 mg
Product Line Tocris
Target Lysine Methyltransferase Inhibitors
Molecular Formula C24H21N5O4