missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UNC 6852

Catalog No. 78165
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78165 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 78165 Supplier Tocris Bioscience™ Supplier No. 7816/5*
Only null left

Polycomb repressive complex 2 (PRC2) Degrader PROTAC™

UNC 6852 is a polycomb repressive complex 2 (PRC2) Degrader (PROTAC™; DC50 = 0.79 μM). It comprises an EED (embryonic ectoderm development) based ligand, joined by a linker to a von Hippel-Lindau ligand. In B lymphoblast cell lines, UNC 6852 selectively degrades PRC2 components EED (DC50 = 0.31 μM, Dmax = 94%), enhancer of zeste homolog 2 (EZH2, DC50 = 0.67 μM, Dmax = 96%), and suppressor of zeste homolog 12 (SUZ12, DC50 = 0.59 μM, Dmax = 82%). In zebrafish, UNC 6852 treatment increases transcription of tfpia and prolongs time to occlusion (TTO) in the caudal vein following laser injury.

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

Specifications

Chemical Name or Material (2 S,4 R)-1-(-2-(4-(4-(5-((Furan-2-ylmethyl)amino)-[1,2,4 ]triazolo[4,3-c]pyrimidin-8-yl)benzamido)butanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide
CAS 2688842-08-0
Quantity 5 mg
Target Active Degraders
Molecular Formula C43H48N10O6S
Purity 0.98