missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ 5M NaCl

Catalog No. A57006
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A57006 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A57006 Supplier Invitrogen™ Supplier No. A57006
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

UltraPure™ 5M NaCl is a filtered, pre-mixed solution containing 292.2 g/L NaCl. This is a common stock solution used in many molecular biology and forensic applications.

  • A filtered, pre-mixed solution containing 292.2 g/L NaCl
  • A common stock solution used in many molecular biology and forensic applications

Specifications

Chemical Name or Material Sodium Chloride
Concentration 5 M
For Use With (Application) Forensics, Molecular Biology
Quantity 4 L
Purity Molecular Biology Grade
Shipping Condition Room Temperature

For Research Use Only.