missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Ultrafree MC Tubes

Catalog No. UFC3000TB
Change view
Click to view available options
For Use With (Application):
Removal of particles and precipitates from aqueous and some solvent-based samples, including protein and nucleic acid solutions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
UFC3000TB Removal of particles and precipitates from aqueous and some solvent-based samples, including protein and nucleic acid solutions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UFC3000TB Supplier MilliporeSigma™ Supplier No. UFC3000TB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Color Clear
Sterility Non-sterile
For Use With (Application) Removal of particles and precipitates from aqueous and some solvent-based samples, including protein and nucleic acid solutions
Housing Material Polypropylene
Max. Force 12,000g (1–4 min.)
Quantity 1000 Pack
Type Filtrate Collection Tube
Volume (Metric) 1.5 mL
Length (Metric) 40 mm
Diameter (Metric) 10.6 mm