Learn More
MilliporeSigma™ Ultrafiltration Discs, 500 kDa NMW, Biomax Polyethersulfone
500 kDa NMWL, Biomax polyethersulfone
Supplier:  MilliporeSigma™ PBVK04310
 View more versions of this product
Catalog No. PBVK04310
Only null left
Description
Description
500 kDa NMWL, Biomax polyethersulfone
Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel™ regenerated cellulose and Biomax™ polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.Features & Benefits:•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm •Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements •Compatible with Amicon™ Stirred Cells Applications:Concentration, Buffer Exchange, Desalting
Specifications
Chemical Identifiers
Ultrafiltration
500 kDa
PES
Ultrafiltration Disc
PES
Protein purification; protein concentration; protein desalting
10Pack
Specifications
Ultrafiltration
500 kDa
13.4 cm2
PES
Ultrafiltration Disc
44.5 mm
PES
PBTK
Protein purification; protein concentration; protein desalting
10Pack
1.75 in.
44.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories