missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Ultrafiltration Discs, 500 kDa NMW, Biomax Polyethersulfone

Catalog No. PBVK02510
Change view
Click to view available options
Diameter (Metric) Filter:
25 mm
44.5 mm
47 mm
63.5 mm
76 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Filter
PBVK02510 25 mm
PBVK04310 44.5 mm
PBVK06210 63.5 mm
PBVK07610 76 mm
PBVK04710 47 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PBVK02510 Supplier MilliporeSigma™ Supplier No. PBVK02510

500 kDa NMWL, Biomax polyethersulfone

Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel™ regenerated cellulose and Biomax™ polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.Features & Benefits:•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm •Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements •Compatible with Amicon™ Stirred Cells Applications:Concentration, Buffer Exchange, Desalting

Specifications

Filter Type Ultrafiltration
Material PES
MWCO 500 kDa
Filter Code PBTK
Filtration Area 4.1 cm2
For Use With (Application) Protein purification; protein concentration; protein desalting
Housing Material PES
Quantity 10Pack
Type Ultrafiltration Disc
Diameter (English) 2.99 in.
Diameter (Metric) 25 mm
Diameter (Metric) Filter 25 mm
Show More Show Less