missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Ultrafiltration Discs, 30 kDa NMW, Ultracel™ Regenerated Cellulose
30 kDa NMWL, Ultracel™ regenerated cellulose
$204.00 - $447.00
Specifications
| Filter Type | Ultrafiltration |
|---|---|
| Material (Membrane) | Regenerated Cellulose |
| MWCO | 30 kDa |
| Filter Code | PLTK |
| For Use With (Application) | Protein purification; protein concentration; protein desalting |
| Catalog Number | Mfr. No. | Diameter (Metric) Filter | Filtration Area | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) Filter | Filtration Area | Price | Quantity | ||||
|
PLTK04310
|
MilliporeSigma™
PLTK04310 |
4.45 cm | 13.4 cm2 |
Pack of 10 Each for $330.00
|
|
||||
|
PLTK07610
|
MilliporeSigma™
PLTK07610 |
7.6 cm | 41.8 cm2 |
Pack of 10 Each for $447.00
|
|
||||
|
PLTK09005
|
MilliporeSigma™
PLTK09005 |
9 cm | 63 cm2 |
Pack of 5 Each for $342.00
|
|
||||
|
PLTK02510
|
MilliporeSigma™
PLTK02510 |
2.5 cm | 4.1 cm2 |
Pack of 10 Each for $204.00
|
|
||||
|
PLTK04710
|
MilliporeSigma™
PLTK04710 |
4.7 cm | 17.3 cm2 |
Pack of 10 Each for $380.00
|
|
||||
|
PLTK06210
|
MilliporeSigma™
PLTK06210 |
6.35 cm | 28.7 cm2 |
Pack of 10 Each for $391.00
|
|
||||
|
PLTK15005
|
MilliporeSigma™
PLTK15005 |
15 cm | 177 cm2 |
Pack of 5 Each for $446.00
|
|
||||
Description
30 kDa NMWL, Ultracel™ regenerated cellulose
Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel™ regenerated cellulose and Biomax™ polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.Features & Benefits:•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm •Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements •Compatible with Amicon™ Stirred Cells Applications:Concentration, Buffer Exchange, DesaltingSpecifications
| Ultrafiltration | |
| 30 kDa | |
| Protein purification; protein concentration; protein desalting | |
| Ultrafiltration Disc |
| Regenerated Cellulose | |
| PLTK | |
| Regenerated Cellulose (RC) |