Learn More
Fisherbrand™ Ultra Low Temperature Recorder
Programmable high and low alarms Features three LEDs (green, yellow and red) to alert user of log-time, temperature warning limit and temperature breach
$659.97
Specifications
| Type | Cryo-Temp Datalogger |
|---|---|
| Data Memory | 32,767 Readings |
| Calibration | Digital Calibration |
| Alarm Types | LED Indicator |
| Battery Type | 3.6 V Lithium |
Alert:
Fire, explosion, and severe burn hazard. Do not recharge,dissassembled, heat above 212°F, incinerate or expose contents to water.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Requires: Interface Package: 15-060-038. Utilizes a USB docking station (sold separately) to communicate with a personal computer.
Description
- Records temperatures as low as -123°F (-86°C)
- Magnetic start
- Thermistor Sensor
- Provides date and time-stamped temperature readings
- Data can be viewed in graphical or tabular form
- Programmable high and low alarms Features three LEDs (green, yellow and red) to alert user of log-time, temperature warning limit and temperature breach
- Material: ABS Plastic, rated IP65
- Battery Life: Two years typical at 25°C, one minute reading intervals
Cryopreservations; Blood Plasma freezers; Vaccines; Cold chain; Low-temperature monitoring; Dry-ice shipment/storage; Shipping and storage applications
Certifications
NIST Traceable
Compliance
CE marked
Specifications
| Cryo-Temp Datalogger | |
| Digital Calibration | |
| 3.6 V Lithium | |
| CE, NIST | |
| Thermistor | |
| 0 to 100% | |
| -122.8°F to +95°F | |
| 1.4 oz. |
| 32,767 Readings | |
| LED Indicator | |
| 2 Years | |
| ABS Plastic | |
| For cryopreservations, Blood plasma freezers, vaccines, cold chain, low temperature monitoring, dry ice shipment/storage, shipping and storage applications | |
| 0.01°C | |
| -86°C to +35°C | |
| 40 g |