missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Ultra Low Temperature Recorder

Catalog No. 15060002
Change view
Click to view available options
Model:
CRYO-TEMP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
15060002 CRYO-TEMP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Fire, explosion, and severe burn hazard. Do not recharge,dissassembled, heat above 212°F, incinerate or expose contents to water.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Catalog No. 15060002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fire, explosion, and severe burn hazard. Do not recharge,dissassembled, heat above 212°F, incinerate or expose contents to water.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Requires

Interface Package: 15-060-038. Utilizes a USB docking station (sold separately) to communicate with a personal computer.

Programmable high and low alarms Features three LEDs (green, yellow and red) to alert user of log-time, temperature warning limit and temperature breach

  • Records temperatures as low as -123°F (-86°C)
  • Magnetic start
  • Thermistor Sensor
  • Provides date and time-stamped temperature readings
  • Data can be viewed in graphical or tabular form
  • Programmable high and low alarms Features three LEDs (green, yellow and red) to alert user of log-time, temperature warning limit and temperature breach
  • Material: ABS Plastic, rated IP65
  • Battery Life: Two years typical at 25°C, one minute reading intervals

Cryopreservations; Blood Plasma freezers; Vaccines; Cold chain; Low-temperature monitoring; Dry-ice shipment/storage; Shipping and storage applications

Certifications

NIST Traceable

Compliance

CE marked

Specifications

Type Cryo-Temp Datalogger
Data Memory 32,767 Readings
Calibration Digital Calibration
Alarm Types LED Indicator
Battery Type 3.6 V Lithium
Battery Life 2 Years
Certifications/Compliance CE, NIST
Material ABS Plastic
Model CRYO-TEMP
Sensor Types Thermistor
For Use With (Application) For cryopreservations, Blood plasma freezers, vaccines, cold chain, low temperature monitoring, dry ice shipment/storage, shipping and storage applications
Humidity Range 0 to 100%
Temperature Resolution 0.01°C
Operating Temperature (English) -122.8°F to +95°F
Operating Temperature (Metric) -86°C to +35°C
Weight (English) 1.4 oz.
Weight (Metric) 40 g
Show More Show Less