Learn More
Promega UDP-Glo Glycosyltransferase Assay + UDP-Glucose, 400 assays
Description
- Universal Glycosyltransferase Activity Assay
- Easy, “add−incubate−read” protocol
- Detects any glycosyltransferase that uses UDP−sugar as a substrate
- Ideal for low−activity glycosyltransferases
Specifications
Specifications
| Content And Storage | Components at different temps |
| Quantity | 400 Assays |