missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ UDP-alpha-D-Galactose, Disodium Salt, >95% Calbiochem™,

Catalog No. 67011110MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67011110MG 10 mg
67011150MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 67011110MG Supplier MilliporeSigma™ Supplier No. 67011110MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A donor substrate for galactosyltransferases used in the biosynthesis of galactose-containing oligosaccharides

Specifications

Molecular Formula C15H22N2Na2O17P2
MDL Number MFCD00077895
Synonym udp-a-d-galactose disodium salt
Solubility Information Easily soluble in cold water
Form Solid
InChI Key PKJQEQVCYGYYMM-UHFFFAOYNA-L
SMILES [Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O
IUPAC Name disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
Molecular Weight (g/mol) 610.27
PubChem CID 126969254
Formula Weight 610.3 g/mol
Percent Purity ≥95%
Quantity 10 mg
Grade HPLC
Chemical Name or Material UDP-alpha-D-Galactose, Disodium Salt
Show More Show Less
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).