missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Series 255 Coveralls

Tyvek coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.

$361.50 - $552.59

Specifications

Product Type Cleanroom Coveralls
Gender Unisex
Material Tyvek Flashspun Polyolefin
Color White
Disposable Single-use
View More Specs

Products 9
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
18-888-101
View Documents
DuPont™
IC255BWHMD00250B
Medium
Case of 25 Each for $361.50
Only null left
 
18-888-102
View Documents
DuPont™
IC255BWHLG00250B
Large
Case of 25 Each for $361.50
Only null left
 
18-888-103
View Documents
DuPont™
IC255BWHXL00250B
X-Large
Case of 25 Each for $361.50
Only null left
 
18-888-104
View Documents
DuPont™
IC255BWH2X00250B
2X-Large
Case of 25 Each for $383.17
Only null left
 
18-888-105
View Documents
DuPont™
IC255BWH3X00250B
3X-Large
Case of 25 Each for $406.15
Only null left
 
18-888-106
View Documents
DuPont™
IC255BWH4X00250B
4X-Large
Case of 25 Each for $438.70
Only null left
 
18-888-107
View Documents
DuPont™
IC255BWH5X00250B
5X-Large
Case of 25 Each for $473.74
Only null left
 
18-888-108
View Documents
DuPont™
IC255BWH6X00250B
6X-Large
Case of 25 Each for $511.64
Only null left
 
18-888-109
View Documents
DuPont™
IC255BWH7X00250B
7X-Large
Case of 25 Each for $552.59
Only null left
 
Description

Description

  • Tyvek™ flashspun polyolefin protective material
  • Patented flash-spinning process
  • Creates barrier to dry particles, microorganisms, non-hazardous liquids
  • Lightweight, durable
  • Serged seams with covered elastic options
  • Traceable
  • Gripper™ soled shoes for slip resistance
  • Bound seams and neck
  • Dolman sleeve
  • Covered elastic wrist, ankle
  • Thumb loop
  • Storm flap
  • Zipper closure
  • White in color
  • Bulk packaged
Specifications

Specifications

Cleanroom Coveralls
Tyvek Flashspun Polyolefin
Single-use
Standard
Bound
CE
Protective Barrier to Dry Particles and Microorganisms
Class 1000 (ISO 6)
Unisex
White
Covered Elastic
Zipper
Non-sterile
Dolman
Bulk
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'