Learn More
DuPont™ Tyvek™ IsoClean™ Series 255 Coveralls
Tyvek coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
Supplier:  DuPont™ IC255BWHXL00250B
 View more versions of this product
Catalog No. 18-888-103
Only null left
Explore available promotions
Description
Description
  • Tyvek™ flashspun polyolefin protective material
  • Patented flash-spinning process
  • Creates barrier to dry particles, microorganisms, non-hazardous liquids
  • Lightweight, durable
  • Serged seams with covered elastic options
  • Traceable
  • Gripper™ soled shoes for slip resistance
  • Bound seams and neck
  • Dolman sleeve
  • Covered elastic wrist, ankle
  • Thumb loop
  • Storm flap
  • Zipper closure
  • White in color
  • Bulk packaged
Specifications
Specifications
Cleanroom Coveralls
Unisex
White
Covered Elastic
Zipper
Non-sterile
Dolman
Bulk
X-Large
Tyvek Flashspun Polyolefin
Single-use
Standard
Bound
CE
Protective Barrier to Dry Particles and Microorganisms
Class 1000 (ISO 6)
Product Suggestions
Product Suggestions