missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Series 255 Coveralls
Tyvek coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
Supplier: DuPont™ IC255BWHLG00250B
Description
- Tyvek™ flashspun polyolefin protective material
- Patented flash-spinning process
- Creates barrier to dry particles, microorganisms, non-hazardous liquids
- Lightweight, durable
- Serged seams with covered elastic options
- Traceable
- Gripper™ soled shoes for slip resistance
- Bound seams and neck
- Dolman sleeve
- Covered elastic wrist, ankle
- Thumb loop
- Storm flap
- Zipper closure
- White in color
- Bulk packaged
Specifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Covered Elastic | |
| Zipper | |
| Non-sterile | |
| Dolman | |
| Bulk |
| Large | |
| Tyvek Flashspun Polyolefin | |
| Single-use | |
| Standard | |
| Bound | |
| CE | |
| Protective Barrier to Dry Particles and Microorganisms | |
| Class 1000 (ISO 6) |