missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrphostin RG 13022 98.0+%, TCI America™
$296.38 - $879.84
Chemical Identifiers
| CAS | 136831-48-6 |
|---|---|
| Molecular Formula | C16H14N2O2 |
| Molecular Weight (g/mol) | 266.3 |
| MDL Number | MFCD00236444 |
| InChI Key | DBGZNJVTHYFQJI-RIYZIHGNSA-N |
| Synonym | 3-(3,4-Dimethoxyphenyl)-2-(pyridin-3-yl)acrylonitrile |
| PubChem CID | 5468216 |
| IUPAC Name | (Z)-3-(3,4-dimethoxyphenyl)-2-pyridin-3-ylprop-2-enenitrile |
| SMILES | COC1=C(C=C(C=C1)C=C(C#N)C2=CN=CC=C2)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T357525MG
|
TCI America
T357525MG |
25 mg |
Each for $296.38
|
|
|||||
|
T3575100MG
|
TCI America
T3575100MG |
100 mg |
Each for $879.84
|
|
|||||
Chemical Identifiers
| 136831-48-6 | |
| 266.3 | |
| DBGZNJVTHYFQJI-RIYZIHGNSA-N | |
| 5468216 | |
| COC1=C(C=C(C=C1)C=C(C#N)C2=CN=CC=C2)OC |
| C16H14N2O2 | |
| MFCD00236444 | |
| 3-(3,4-Dimethoxyphenyl)-2-(pyridin-3-yl)acrylonitrile | |
| (Z)-3-(3,4-dimethoxyphenyl)-2-pyridin-3-ylprop-2-enenitrile |
Specifications
| 136831-48-6 | |
| Yellow | |
| MFCD00236444 | |
| 3-(3,4-Dimethoxyphenyl)-2-(pyridin-3-yl)acrylonitrile | |
| COC1=C(C=C(C=C1)C=C(C#N)C2=CN=CC=C2)OC | |
| 266.3 | |
| 266.30 | |
| Crystalline Powder |
| 118°C | |
| C16H14N2O2 | |
| 25 mg | |
| DBGZNJVTHYFQJI-RIYZIHGNSA-N | |
| (Z)-3-(3,4-dimethoxyphenyl)-2-pyridin-3-ylprop-2-enenitrile | |
| 5468216 | |
| ≥98.0% (GC) | |
| Tyrphostin RG 13022 |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator