Learn More
DuPont™ Tychem™ 2000 Coverall (all)
Flexible and durable light liquid splash protection
$162.91 - $232.70
Specifications
Gender Unisex
Color Yellow
Ankle Style Elastic
View More Specs
Products 18
Catalog Number Mfr. No. Seam Type Size Price Quantity  
Catalog Number Mfr. No. Seam Type Size Price Quantity  
19-162-759
View Documents
DuPont™
QC125TYL5X000400
Taped 5X-Large
Case of 4 Each for $213.55
Only null left
18-178C
View Documents
DuPont™
QC125SYLLG001200
Serged Large
Case of 12 Each for $183.37
Only null left
18178F
View Documents
DuPont™
QC125SYL3X001200
Serged 3X-Large
Case of 12 Each for $189.45
Only null left
19-162-757
View Documents
DuPont™
QC125TYL3X000400
Taped 3X-Large
Case of 4 Each for $183.08
Only null left
19-162-761
View Documents
DuPont™
QC125TYLMD000400
Taped Medium
Case of 4 Each for $162.91
Only null left
19-162-760
View Documents
DuPont™
QC125TYLLG000400
Taped Large
Case of 4 Each for $162.91
Only null left
18-178D
View Documents
DuPont™
QC125SYLXL001200
Serged X-Large
Case of 12 Each for $168.61
Only null left
19-162-758
View Documents
DuPont™
QC125TYL4X000400
Taped 4X-Large
Case of 4 Each for $197.74
Only null left
19-053-921
View Documents
DuPont™
QC125BYL2X001200
Bound 2X-Large
Case of 12 Each for $232.70
Only null left
18178E
View Documents
DuPont™
QC125SYL2X001200
Serged 2X-Large
Case of 12 Each for $178.77
Only null left
19017977B
View Documents
DuPont™
QC125BYLMD001200
Bound Medium
Case of 12 Each for $219.54
Only null left
191202582
View Documents
DuPont™
QC125SYL5X001200
Serged 5X-Large
Case of 12 Each for $220.98
Only null left
19-017-975
View Documents
DuPont™
QC125BYLLG001200
Bound Large
Case of 12 Each for $219.54
Only null left
18-178B
View Documents
DuPont™
QC125SYLMD001200
Serged Medium
Case of 12 Each for $168.61
Only null left
19-162-762
View Documents
DuPont™
QC125TYLXL000400
Taped X-Large
Case of 4 Each for $162.91
Only null left
19-162-756
View Documents
DuPont™
QC125TYL2X000400
Taped 2X-Large
Case of 4 Each for $172.71
Only null left
19-017-976
View Documents
DuPont™
QC125BYLXL001200
Bound X-Large
Case of 12 Each for $219.54
Only null left
19-067-017
View Documents
DuPont™
QC125SYL4X001200
Serged 4X-Large
Case of 12 Each for $204.59
Only null left
Description
Description
Tychem™ QC is polyethylene-coated Tyvek™ fabric fabric provides at least 30 minutes of protection against 42 chemical challenges
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Yellow for high visibility
  • 10 mil thickness
QC 125B
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
QC 125S
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
QC 125T
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
Compliance
Meets ASTM™ D3776, ASTM D1777, ASTM 1117, ASTM D3786, ASTM D5034 standards.
Specifications
Specifications
Unisex
Yellow
Elastic
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'