missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Two Compartment Petri Dish
For comparative measurements/conditions or use with different agar systems
Supplier: Greiner Bio-One 635102
Description
Manufactured from crystal clear and heat-resistant polystyrene for analysis by microscopySpecifications
| 25 mL | |
| Polystyrene | |
| Non-sterile | |
| 480 | |
| Yes | |
| 94 mm |
| Clear | |
| Round | |
| 2 | |
| 20 | |
| 15 mm |