missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tween 20 washing buffer, 1% in PBS (10X)

$138.86

Specifications

Chemical Name or Material Buffer
Concentration 1% in PBS (10X)
Physical Form Viscous Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ63314AP
View Documents Promotion Details
Thermo Scientific Chemicals
J63314AP
500 mL
Each for $138.86
Only null left
 
Description

Description

Tween-20 is a non-ionic surfactant that is useful for solubilizing membrane proteins during isolation. Tween-20 is also added to buffers for immunohistochemistry. Tween-20 may also help to prevent non-specific antibody binding and function as a blocking agent in immunoblotting assays. It has been used in preextraction of membranes to remove peripheral proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Buffer
1% in PBS (10X)
Viscous Liquid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only