missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween 20 (=Polyoxyethylene Sorbitan Monolaurate), TCI America™
$32.28 - $47.46
Chemical Identifiers
| CAS | 9005-64-5 |
|---|---|
| Molecular Formula | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 |
| Molecular Weight (g/mol) | 522.68 |
| MDL Number | MFCD00165986 |
| InChI Key | HMFKFHLTUCJZJO-UHFFFAOYNA-N |
| Synonym | Polyoxyethylene Sorbitan Monolaurate |
| PubChem CID | 443314 |
| IUPAC Name | 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
| SMILES | CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Chemical Identifiers
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
| MFCD00165986 | |
| Polyoxyethylene Sorbitan Monolaurate | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
Specifications
| Yellow | |
| Clear Liquid at 20°C | |
| 6 to 7.5 (5% H2O soln.) | |
| MFCD00165986 | |
| HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate | |
| 443314 |
| 9005-64-5 | |
| -14°C | |
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
| Polyoxyethylene Sorbitan Monolaurate | |
| CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 522.68 | |
| Tween 20 (=Polyoxyethylene Sorbitan Monolaurate) |
Safety and Handling
EINECSNumber : (8)-0055
RTECSNumber : TR7400000
TSCA : Yes