missing translation for 'onlineSavingsMsg'
Learn More

Tween 20 (=Polyoxyethylene Sorbitan Monolaurate), TCI America™

Catalog No. T054325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T054325G 25 g
T0543500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T054325G Supplier TCI America Supplier No. T054325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyoxyethylene Sorbitan Monolaurate
PubChem CID 443314
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow
Physical Form Clear Liquid at 20°C
Melting Point -14°C
pH 6 to 7.5 (5% H2O soln.)
Quantity 25 g
Chemical Name or Material Tween 20 (=Polyoxyethylene Sorbitan Monolaurate)
EINECSNumber (8)-0055
RTECSNumber TR7400000
TSCA Yes