missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tween™ 20 Surfact-Amps™ Detergent Solution

Catalog No. p-6014006
Change view
Click to view available options
Quantity:
50 mL
250 mL
500 mL
6 x 10 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI85113 50 mL
PI28320 6 x 10 mL
PI85114 250 mL
PI85115 500 mL
PI28321 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PI85113 Supplier Thermo Scientific™ Supplier No. 85113
Only null left

Thermo Scientific™ Surfact-Amps 20 Detergent Solution is highly-purified Tween-20 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.

Thermo Scientific™ Tween-20 Detergent Surfact-Amps Solution is highly-purified Tween-20 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.

This is an easy-to-use 10% (w/v) solution of purified Tween-20 detergent for use in routine and high-demand protein research methods and molecular biology techniques. Tween-20 is a nonionic polyoxyethylene surfactant that is most frequently added to PBS or TBS wash buffers for ELISA, Western blotting and other immunoassay methods.

Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Highlights:

Tween 20 – a popular nonionic detergent for use in immunoassay wash buffers

Accurate – precise 10% detergent solution in ultrapure water

Easy-to-use – solution is simple to dispense and dilute for use

Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls

Stable – packed in glass ampules under inert nitrogen gas

Detergent Properties:

  • Molecular Weight: 1228
  • Detergent Class: Nonionic
  • Aggregation Number: Unknown
  • Micelle Molecular Weight: Unknown
  • Critical Micelle Concentration (CMC): 0.06mM (0.0074%, w/v)
  • Cloud Point: 95°C
  • Dialyzable: No

 

Specifications

Product Type Detergent
Content And Storage Store in a cool, dry, and well-ventilated area
Form Liquid
Product Line Surfact-Amps, Tween
Reagent Type Detergent Solution
Quantity 50 mL

For Research Use Only. Not for use in diagnostic procedures.