missing translation for 'onlineSavingsMsg'
Learn More

Turbidity Stock Solution 2, 10% (w/v) Hexamine, Ricca Chemical

$117.70 - $117.70

Specifications

Chemical Name or Material Turbidity Stock Solution 2
Packaging Amber Glass Bottle
CAS 100-97-0 , 7732-18-5
CAS Min % 89.0,8.91
Concentration or Composition Notes 10% (w/v) Hexamine
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
88324
View Documents
Ricca Chemical Company
88324
120 mL
Each for $117.70
Only null left
 
Description

Description

Specifications

Specifications

Turbidity Stock Solution 2
100-97-0 , 7732-18-5
10% (w/v) Hexamine
Liquid
Approximately 0°C
Amber Glass Bottle
89.0,8.91
93.0,9.28
Colorless
Videos
SDS
Documents

Documents