missing translation for 'onlineSavingsMsg'
Learn More

Turbidity Standard, 500 NTU, Formazin, Ricca Chemical

Catalog No. R8819500500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R8819500500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R8819500500 Supplier Ricca Chemical Company Supplier No. R8819500500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-97-0 , 10034-93-2 , 7732-18-5
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
PubChem CID 4101
ChEBI CHEBI:6824
SMILES C1N2CN3CN1CN(C2)C3

Specifications

Packaging Amber Glass Bottle
Odor Odorless
Assay Percent Range 500 ± 5 NTU
Physical Form Suspension
Color White
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Infinitely soluble in water
Solution Type Calibration Standard
Type Analytical Standard
Solvent or Matrix Water
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Turbity Standard, 500 NTU (Nephelometric Turbidity Units) - Formazin
Show More Show Less