missing translation for 'onlineSavingsMsg'
Learn More

Turbidity Standard, 4000 NTU, LabChem™

Supplier:  LabChem LC262901

Catalog No. LC262901


Only null left

Description

Description

Specifications

Specifications

Turbidity Standard
Poly Bottle
94.5,5,0.5
Liquid
H4N2·H2SO4
Formazin
VKYKSIONXSXAKP-UHFFFAOYSA-N
140.19
4101
130.12
4000 NTU
100-97-0 , 10034-93-2 , 7732-18-5
Passes Test
C6H12N4
Colorless
Soluble in water
C1N2CN3CN1CN(C2)C3
500 mL
CHEBI:6824

Chemical Identifiers

100-97-0
140.19
Formazin
CHEBI:6824
C6H12N4
VKYKSIONXSXAKP-UHFFFAOYSA-N
4101
C1N2CN3CN1CN(C2)C3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
May cause cancer.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist, vapors, spray.
Wear eye protection, protective gloves.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature; Mix well before use

SDS
Documents

Documents

Promotions

Promotions