missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten zirconium oxide, 99.7% (metals basis excluding Hf), Hf <0.38%, Thermo Scientific Chemicals
$587.85 - $2376.10
Chemical Identifiers
| CAS | 16853-74-0 |
|---|---|
| MDL Number | MFCD00168096 |
| Synonym | Zirconium tungstate; Zirconium tungsten oxide |
Description
Tungsten zirconium oxide is used as a super acid catalyst in the hydrocarbon isomerization process and esterification of waste acid oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 16853-74-0 | |
| MFCD00168096 | |
| Zirconium tungstate; Zirconium tungsten oxide |
Specifications
| Green to White | |
| Powder | |
| MFCD00168096 | |
| Insoluble in water. | |
| 99.7% | |
| 5.10 g/mL | |
| (metals basis excluding Hf), Hf <0.3% |
| 16853-74-0 | |
| 50 g | |
| Zirconium tungstate; Zirconium tungsten oxide | |
| 586.92 | |
| Odorless | |
| Tungsten zirconium oxide |
Safety and Handling
EINECSNumber : 240-876-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only