missing translation for 'onlineSavingsMsg'
Learn More

Tungsten zirconium oxide, 99.7% (metals basis excluding Hf), Hf <0.38%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04137518

 View more versions of this product

Catalog No. AA4137518


Only null left
Explore available promotions

Description

Description

Tungsten zirconium oxide is used as a super acid catalyst in the hydrocarbon isomerization process and esterification of waste acid oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tungsten zirconium oxide
16853-74-0
50 g
MFCD00168096
Insoluble in water.
99.7%
5.10 g/mL
Green to White
Powder
(metals basis excluding Hf), Hf <0.3%
Zirconium tungstate; Zirconium tungsten oxide
586.92
Odorless

Chemical Identifiers

16853-74-0
MFCD00168096
Zirconium tungstate; Zirconium tungsten oxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 240-876-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only