missing translation for 'onlineSavingsMsg'
Learn More

Tungsten foil, 0.5mm (0.02in) thick, 99.95% (metals basis)

Form: Approx. 12g/25 x 50mm | CAS: 7440-33-7 | W | 183.84 g/mol

$231.26 - $2676.21

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10414FG
View Documents
Thermo Scientific Chemicals
010414FG
25 x 50 mm
Each for $231.26
Only null left
 
AA10414FY
View Documents
Thermo Scientific Chemicals
010414FY
50 x 100 mm
Each for $315.99
Only null left
 
AA10414GD
View Documents
Thermo Scientific Chemicals
010414GD
50 x 500 mm
Each for $1,006.22
Only null left
 
AA10414GW
View Documents
Thermo Scientific Chemicals
010414GW
200 x 200 mm
Each for $1,406.95
Only null left
 
AA10414HB
View Documents
Thermo Scientific Chemicals
010414HB
300 x 300 mm
Each for $2,676.21
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]

Specifications

Silver to Gray
Silver Gray foil
99.95% (Metals basis)
W
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
7440-33-7
25 x 50 mm
Foil
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Tungsten
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-143-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only