Learn More
Tungsten foil, 0.5mm (0.02in) thick, 99.95% (metals basis)
Form: Approx. 12g/25 x 50mm | CAS: 7440-33-7 | W | 183.84 g/mol
Supplier: Thermo Scientific Chemicals 010414GW
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver to Gray | |
| 7440-33-7 | |
| 200 x 200 mm | |
| Foil | |
| MFCD00011461 | |
| WFKWXMTUELFFGS-UHFFFAOYSA-N | |
| tungsten | |
| 23964 |
| Tungsten | |
| Silver Gray foil | |
| 99.95% (Metals basis) | |
| W | |
| wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
| [W] | |
| 183.84 | |
| CHEBI:27998 |
Chemical Identifiers
| 7440-33-7 | |
| 183.84 | |
| WFKWXMTUELFFGS-UHFFFAOYSA-N | |
| 23964 | |
| tungsten |
| W | |
| MFCD00011461 | |
| wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
| CHEBI:27998 | |
| [W] |
Safety and Handling
EINECSNumber : 231-143-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only