missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ SuperSW2000 HPLC Column, 4 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021965
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021965 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021965 Supplier Merck Emd Millipore Supplier No. 818674

Featuring ultra-efficient 4 μm particles, TSKgel SuperSW2000 provides higher efficiency than 5 μm TSKgel G2000SWxl Columns. The 125 Å pore size results in a fractionation range up to 150,000 Da for globular proteins.

  • The narrow 4.6 mm column inner diameter reduces buffer consumption and is ideal for sample-limited applications
  • To fully benefit from the high efficiency of 4 μm columns, and because the column diameter of TSKgel SuperSW columns is 4.6 mm, it is important to minimize the amount of dead volume in the HPLC system
  • Salt Concentration: ≤ 0.5 M

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 0.4 mL/min
Length (Metric) 30 cm
Max. Pressure 120 bar
Particle Size 4 μm
pH 2.5 to 7.5
Pore Size 125 Å
Solvents 0-100% Aqueous Soluble Organic Solvents
Temperature 10°C to 30°C
USP Type L20, L59
Stationary Phase Diol
Product Line TSKgel
Shipping Condition Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M Phosphate Buffer
Matrix Diol Phase/Silica Particle Platform
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation