missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ SuperSW3000 HPLC Column, 4 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022055
Change view
Click to view available options
Diameter (Metric):
2 mm
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
111022055 2 mm
111021966 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022055 Supplier Merck Emd Millipore Supplier No. 821485

Featuring ultra-efficient 4 μm particles, TSKgel SuperSW3000 provides higher efficiency than 5 μm TSKgel G2000SWxl Columns. The 125 Å pore size results in a fractionation range up to 150,000 Da for globular proteins.

  • The narrow 4.6 mm column inner diameter reduces buffer consumption and is ideal for sample-limited applications
  • To fully benefit from the high efficiency of 4 μm columns, and because the column diameter of TSKgel SuperSW columns is 4.6 mm, it is important to minimize the amount of dead volume in the HPLC system
  • Salt Concentration: ≤ 0.5 M

Specifications

Diameter (Metric) 2 mm
Flow Rate 0.075 mL/min
Length (Metric) 30 cm
Max. Pressure 120 bar
Particle Size 4 μm
pH 2.5 to 7.5
Pore Size 250 Å
Solvents 0-100% Aqueous Soluble Organic Solvents
Temperature 10°C to 30°C
USP Type L20, L59
Stationary Phase Diol
Product Line TSKgel
Shipping Condition Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M Phosphate Buffer
Matrix Diol Phase/Silica Particle Platform
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation