missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ SuperHZM-H HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022018
Change view
Click to view available options
Diameter (Metric):
4.6 mm
6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
111022018 4.6 mm
111022019 6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022018 Supplier Merck Emd Millipore Supplier No. 819664

TSKgel™ SuperHZM-H Mixed-Bed Columns are packed with rigid, porous, polymer beads each having a varying mean pore diameter. They are used for the non-aqueous separation of organic-soluble polymers with high molecular weights of less than 400,000,000 Da.

  • THF shipping solvent may be replaced by a limited number of organic solvents

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 0.4 mL/min
Length (Metric) 15 cm
Max. Pressure 10 bar
Particle Size 10 μm
Temperature 80°C
Stationary Phase Polystyrene-divinylbenzene
Product Line TSKgel
Material Stainless Steel Hardware
Technique SEC

TSKgel is a registered trademark of Tosoh Corporation