missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ SuperHZM-H Guard Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022022
Change view
Click to view available options
Length (Metric):
2 cm
3.5 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111022022 2 cm
111022021 3.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022022 Supplier Merck Emd Millipore Supplier No. 819668

TSKgel™ SuperHZM-H Mixed-Bed Columns are packed with rigid, porous, polymer beads each having a varying mean pore diameter. They are used for the non-aqueous separation of organic-soluble polymers with high molecular weights of less than 400,000,000 Da.

  • THF shipping solvent may be replaced by a limited number of organic solvents

Specifications

Particle Size 10 μm
Length (Metric) 2 cm
Diameter (Metric) Inner 4.6 mm
Flow Rate 0.4 mL/min
Stationary Phase Polystyrene-divinylbenzene
Temperature (Metric) Maximum 80°C
Material Stainless Steel
Technique Size Exclusion (SEC)
Product Line TSKgel

TSKgel is a registered trademark of Tosoh Corporation