missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ SuperH4000 HPLC Column, 3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021912
Change view
Click to view available options
Length (Metric):
15 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021912 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021912 Supplier Merck Emd Millipore Supplier No. 817994

Packed with rigid, porous polymer beads and used for the non-aqueous separation of polymers with molecular weights less than 400,000 Da.

  • A high efficiency/high throughput version of the conventional TSKgel HHR Column
  • Both column types are based on the same bead chemistry
  • High-throughput gel permeation columns have dimensions of 6 mm ID x 15 cm
  • Shipped in THF, which can be exchanged for a wide variety of organic solvents
  • Maximum Operating Temperature: 140°C

Specifications

Diameter (Metric) 6 mm
Flow Rate 0.8 mL/min
Length (Metric) 15 cm
Max. Pressure 40 bar
Particle Size 3 μm
Pore Size 200 Å
Temperature 140°C
USP Type L21
Stationary Phase Polystyrene-divinylbenzene
Product Line TSKgel
Shipping Condition Shipped in THF
Matrix Polystyrene-divinylbenzene Phase/Polymer Particle Platform
Material Stainless Steel Hardware
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation