missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ Super-Phenyl HPLC Column, 2.3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022038
Change view
Click to view available options
Length (Metric):
5 cm
10 cm
Diameter (Metric):
2 mm
4.6 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
111022038 10 cm 2 mm
111022037 5 cm 2 mm
111021937 5 cm 4.6 mm
111021938 10 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111022038 Supplier Merck Emd Millipore Supplier No. 820018

TSKgel™ Super-Phenyl Columns are packed with monodispersed 2 μm (nominal, mean particle size is 2.3 μm) spherical silica particles covalently bonded with phenyl groups.

  • The small particle size used in Super Series and TSKgel ODS-140HTP Columns provide the highest efficiency of all TSKgel reversed phase columns
  • Monodispersity of the particles reduces column back pressure to values more typical of larger particles, allowing the use of higher flow rates compared to other 2 μm packings
  • TSKgel Super-Phenyl Columns are less hydrophobic than TSKgel Super-Octyl and TSKgel Super-ODS Columns
  • An excellent choice for peptides, proteins, amino acids, tryptic digests, nucleotides, pharmaceutical molecules and food/beverage samples

Specifications

Diameter (Metric) 2 mm
Flow Rate 0.25 mL/min
Length (Metric) 10 cm
Max. Pressure 150 bar
Particle Size 2.3 μm
pH 2 to 7.5
Solvents 0-100% Organic Solvent
Surface Area 100 m2/g
Temperature 10°C to 50°C
Stationary Phase Phenyl
Product Line TSKgel
Shipping Condition Shipped in 50% Acetonitrile, 50% Water
Matrix Phenyl Phase/Silica Particle Platform
Carbon Load 3%
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation