missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ SP-STAT HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022094
Change view
Click to view available options
Length (Metric):
3.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111022094 3.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022094 Supplier Merck Emd Millipore Supplier No. 821963

TSKgel™™ SP-STAT columns are packed with hydrophilic non-porous resin particles of which the surface consists of an open access network of multi-layered cation exchange groups.

TSKgel™™ SP-STAT's innovative bonding chemistry, combined with a relatively large particle size, results in a respectable loading capacity and a low operating pressure. Applications for the TSKgel™ SP-STAT columns include the separation of proteins, protein aggregates, charge isomers of monoclonal antibodies, PEGylated proteins, and peptide digests.

Order Info

Shipped in DI water.

Specifications

Diameter (Metric) 3 mm
Flow Rate 1 to 2 mL/min.
Length (Metric) 3.5 cm
Max. Pressure 100 bar
Particle Size 10 μm
pH 3.0 - 10
Solvents ≤50% organic solvent
Temperature 10°C to 60°C
Stationary Phase Sulfonic Acid
Product Line TSKgel
Matrix Polymer Particle Platform
Phase Cation Exchange
Material Stainless Steel Hardware
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation