missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ SP-5PW HPLC Column, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021828
Change view
Click to view available options
Diameter (Metric):
2 mm
5 mm
8 mm
20 mm
Material:
Glass
Stainless Steel Hardware
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Material
111021828 5 mm Glass
111021822 8 mm Glass
111021847 20 mm Glass
111021971 2 mm Stainless Steel Hardware
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111021828 Supplier Merck Emd Millipore Supplier No. 813062

For the separation and analysis of proteins, peptides, and other biologically active molecules.

  • Packed with porous hydrophilic polymer beads that are surface modified with a strong cation exchanger
  • For the separation and analysis of proteins, peptides, and other biologically active molecules
  • Columns have the same backbone chemistry and selectivity as the bulk process scale resins Toyopearl SP-650 & TSKgel SP-5PW (20) and (30)
  • Salt Concentration: ≤1 M

Specifications

Diameter (Metric) 5 mm
Flow Rate 1 mL/min
Ion Exchange Group Na+
Length (Metric) 5 cm
Max. Pressure 150 bar
Particle Size 10 μm
pH 2 to 12
Solvents ≤ 20% Organic Solvent
Temperature 10°C to 45°C
Stationary Phase Sulfonic Acid
Product Line TSKgel
Matrix Sulfonic Acid Phase/Hydroxylated Methacrylic Polymer
Material Glass
Technique Cation Exchange
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation