missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ QC-PAK GFC 300 HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021858
Change view
Click to view available options
Length (Metric):
15 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021858 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021858 Supplier Merck Emd Millipore Supplier No. 816049

These columns are 15 cm in length with 5 μm particles and offer the same resolution in half the time as the 30 cm, 10 μm TSKgel G2000SW and G3000SW columns. Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M Phosphate Buffer, pH 6.7.

  • Salt Concentration: ≤ 0.5 M

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 1.2 mL/min
Length (Metric) 15 cm
Max. Pressure 41 bar
Particle Size 5 μm
pH 2.5 to 7.5
Pore Size 250 Å
Solvents 0-100% Aqueous Soluble Organic Solvents
Temperature 10°C to 30°C
USP Type L20, L59
Stationary Phase Diol
Product Line TSKgel
Shipping Condition Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M Phosphate Buffer
Matrix Diol Phase/Silica Particle Platform
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation