missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ QC-PAK GFC 200 HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021873
Change view
Click to view available options
Length (Metric):
15 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021873 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021873 Supplier Merck Emd Millipore Supplier No. 816215

Widely considered the premium columns for analyzing biopolymers by GFC, TSKgel G2000SWXL Columns feature high pore volume per unit column volume, low sample adsorption, and excellent column efficiency.

  • With 5 μm particles and 125 Å pores, TSKgel G2000SWXL is an excellent choice for protein and large peptide separations
  • The fractionation range is applicable for globular protein samples up to 150,000 Da.
  • Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M Phosphate Buffer, pH 6.7
  • Salt Concentration: ≤ 0.5 M

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 1.2 mL/min
Length (Metric) 15 cm
Max. Pressure 41 bar
Particle Size 5 μm
pH 2.5 to 7.5
Pore Size 125 Å
Solvents 0-100% Aqueous Soluble Organic Solvents
Temperature 10°C to 30°C
USP Type L20, L59
Stationary Phase Diol
Product Line TSKgel
Shipping Condition Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M Phosphate Buffer
Matrix Diol Phase/Silica Particle Platform
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation