missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ Protein C4-300 Guard Cartridge, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022188
Change view
Click to view available options
Diameter (Metric) Inner:
2 mm
3.2 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
111022188 3.2 mm
111022189 2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022188 Supplier Merck Emd Millipore Supplier No. 822833

Addition to the extensive line of Reversed Phase columns from Tosoh Bioscience

  • The high efficiency columns are designed for the optimal recovery and resolution of proteins such as recombinant proteins, antibody fragments or PEGylated proteins.
  • The 300 Å pore size of the TSKgel Protein C4-300 columns are ideal for the separation of proteins.
  • A particle size of 3 μm and optimized ligand density and alkyl length result in better protein and peptide resolution compared to other leading RP-C4 HPLC phases.
  • Latest surface modification techniques and endcapping of residual silanol groups reduce undesirable secondary interactions and peak tailing.
  • Shipped in acetonitrile.
  • 3% Carbon, endcapped with trimethylsilane
  • No limitation salt concentration
  • 0-100% organic solvent

Specifications

pH 1.5 - 7.5
Particle Size 3 μm
Length (Metric) 1.5 cm
Surface Area 100 m2/g
Diameter (Metric) Inner 3.2 mm
Flow Rate 1 mL/min
For Use With (Equipment) L26 Per USP
Stationary Phase C4 (butyl)
Temperature (Metric) Maximum 10°C to 50°C
Technique Reversed phase
Matrix Spherical silica
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation