missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ ODS-140HTP HPLC Column, 2.3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022086
Change view
Click to view available options
Length (Metric):
5 cm
10 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111022086 5 cm
111022087 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022086 Supplier Merck Emd Millipore Supplier No. 821927
Only null left

Provides high resolution and short analyses times at moderate pressures, enabling high throughput separations.

  • Polylayer bonding chemistry of the 2.3 μm particle size columns results in highly efficient and durable columns
  • The lower pressure drop reduces the burden on the hardware, allowing the TSKgel ODS-140HTP Columns to be used with either UHPLC (up to 9000 psi) or conventional HPLC systems
  • For use in high throughput applications including drug discovery, pharmacokinetics and peptide digest separations
  • Excellent peak shape for basic compounds

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 5 cm
Max. Pressure 600 bar
Particle Size 2.3 μm
pH 2 to 7.5
Solvents 0-100% Organic Solvent
Surface Area 100 m2/g
Temperature 10°C to 50°C
USP Type L1
Stationary Phase C18 (octadecyl)
Product Line TSKgel
Shipping Condition Shipped in Acetonitrile
Matrix C18 (octadecyl) Phase/Spherical Silica
Carbon Load 8%
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation