missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSKgel™ Octadecyl-2PW HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™
Highly cross-linked polymethacrylate base material provides excellent stability in high pH buffer systems. Withstands rigorous cleaning with either acid or base, 125 Å pores allow for analysis of peptides up to 8,000 Da.
$1561.85 - $2323.30
Specifications
| Length (Metric) | 15 cm |
|---|---|
| Ligand Type | 1 meq/mL (Estimated Ligand Density) |
| Particle Size | 5 μm |
| Pore Size | 125 Å |
| Solvents | 0-100% Organic Solvent |
Description
- Columns packed with larger particle sizes are available for semi-preparative scale separations
- Polymer-based particles for reversed-phase (RP) HPLC are more pH tolerant, and prevent silanol interactions that are common with silica-based particles
- Some silicas have an intrinsic metal content that may cause chelation problems with sensitive biomolecules, leading to poor recovery
- Polymeric TSKgel Columns come in C18 and phenyl functional groups, as well as a range of particle porosities
- Fast kinetics and quantitative protein recovery at submicrogram loading are characteristic of nonporous, resin-based TSKgel Octadecyl-NPR Columns
- Shipped in 55% Acetonitrile, 45% Water
Specifications
| 15 cm | |
| 5 μm | |
| 0-100% Organic Solvent | |
| C18 (octadecyl) | |
| C18 (octadecyl) Phase/Polymer Particle Platform |
| 1 meq/mL (Estimated Ligand Density) | |
| 125 Å | |
| 4°C to 50°C | |
| Shipped in 55% Acetonitrile, 45% Water | |
| Stainless Steel Hardware |
TSKgel is a registered trademark of Tosoh Corporation