missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ GMHHR-M HPLC Column, 5 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021902
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021902 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021902 Supplier Merck Emd Millipore Supplier No. 817392

TSKgel GMHHR-M Mixed Bed Columns are packed with rigid, multiple pore size, polymer beads. Mixed bed columns are commonly used for unknown samples and for the analysis of polymers with a broad molecular weight distribution.

  • Conventional GPC Columns with dimensions of 7.8 mm ID x 30 cm
  • TSKgel GMHHR-M Columns are available in 5 and 13 μm particle sizes
  • If shearing occurs when using small (5 μm) particle size columns, use of larger (13 μm) columns is recommended
  • All TSKgel HHR-type columns are shipped in THF, which can be exchanged for a broad range of organic solvents
  • Maximum Operating Temperature: 140°C

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 0.5 to 1.0 mL/min
Length (Metric) 30 cm
Max. Pressure 50 bar
Particle Size 5 μm
Temperature 80°C
USP Type L21
Stationary Phase Polystyrene-divinylbenzene
Product Line TSKgel
Shipping Condition Shipped in THF
Matrix Polystyrene-divinylbenzene Phase/Polymer Particle Platform
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation