missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ GMHHR-H(S) Guard Column, 13 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021898
Change view
Click to view available options
7.5 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111021898 7.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021898 Supplier Merck Emd Millipore Supplier No. 817367

Guard Column for use with TSKgel™ GMHHR-H(S) HPLC Columns (817361). TSKgel™ GMHHR-H mixed bed columns are packed with rigid, multiple pore size, polymer beads.

  • Conventional GPC columns with dimensions of 7.8 mm ID x 30 cm
  • Mixed bed columns are commonly used for unknown samples and for the analysis of polymers with a broad molecular weight distribution
  • All TSKgel HHR-type columns are shipped in THF, which can be exchanged for a broad range of organic solvents
  • Maximum Operating Temperature: 140°C

Specifications

Particle Size 13 μm
Length (Metric) 7.5 cm
Diameter (Metric) Inner 7.5 mm
Flow Rate 0.5 to 1.0 mL/min
Stationary Phase Polystyrene-divinylbenzene
Temperature (Metric) Maximum 80°C
Material Stainless Steel
Technique Size Exclusion (SEC)
USP Type L21
Product Line TSKgel

TSKgel is a registered trademark of Tosoh Corporation