missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ GMHHR-H HPLC Columns, 20 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021956
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111021956 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111021956 Supplier Merck Emd Millipore Supplier No. SGMA818399

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TSKgel™ GMHhr-H Mixed Bed Columns are packed with rigid, multiple pore size, polymer beads. There columns are commonly used for unknown samples, and for the analysis of polymers with a broad molecular weight distribution.

  • Conventional GPC columns with dimensions of 7.8 mm ID x 30 cm
  • Columns are shipped in THF, which can be exchanged for a wide variety of organic solvents
  • TSKgel GMHhr-H (13, 20 or 30 μm) are recommended to prevent or limit shearing of ultra high molecular weight polymers, if this occurs when using 5 μm particle size columns
  • Maximum Operating Temperature: 80°C

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 0.5 to 1 mL/min
Length (Metric) 30 cm
Max. Pressure 15 bar
Particle Size 20 μm
Temperature 80°C
USP Type L21
Stationary Phase Polystyrene-divinylbenzene
Product Line TSKgel
Shipping Condition Shipped in THF
Material Stainless Steel
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation